CO1H324A67BA CombiPhos Catalysts Inc

4-Fluoro-2-(diethylamino)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223048-57-3
  • Molecular Weight : 294.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart