CO1H324A67BD CombiPhos Catalysts Inc

4-Fluoro-2-(3-methyl-1H-pyrazol-1-yl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223035-08-1
  • Molecular Weight : 303.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart