CO1H324A67C0 CombiPhos Catalysts Inc

4-Fluoro-2-(2-methylimidazol-1-yl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223049-13-4
  • Molecular Weight : 303.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart