CO1H324A67C9 CombiPhos Catalysts Inc

4-Fluoro-2-(thiazol-5-yl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223034-03-3
  • Molecular Weight : 306.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart