CO1H324A67CF CombiPhos Catalysts Inc

5-Fluoro-2-(thiazol-2-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223049-18-9
  • Molecular Weight : 306.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart