CO1H324A67D3 CombiPhos Catalysts Inc

5-Fluoro-2-(1H-pyrazol-1-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223045-29-0
  • Molecular Weight : 289.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart