CO1H324A67D7 CombiPhos Catalysts Inc

5-Fluoro-2-(2-methylimidazol-1-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223034-88-4
  • Molecular Weight : 303.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart