CO1H324A67DC CombiPhos Catalysts Inc

5-Fluoro-2-(3-methylpiperidin-1-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2222997-16-0
  • Molecular Weight : 320.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart