CO1H324A67DE CombiPhos Catalysts Inc

5-Fluoro-2-(morpholino)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223048-20-0
  • Molecular Weight : 308.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart