CO1H324A67F5 CombiPhos Catalysts Inc

2-fluoro-6-isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 2223048-99-3
  • Molecular Weight : 265.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart