CO1H324A6810 CombiPhos Catalysts Inc

2-(Cyclohexyl)-6-(oxolan-3-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223004-35-9
  • Molecular Weight : 357.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart