CO1H324A6814 CombiPhos Catalysts Inc

2-(Oxan-4-yl)-6-(oxolan-3-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2222997-40-0
  • Molecular Weight : 359.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart