CO1H324A6825 CombiPhos Catalysts Inc

2-(iso-Butyl)-6-(N-Boc-piperidin-3-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2222995-97-1
  • Molecular Weight : 444.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart