CO1H324A6836 CombiPhos Catalysts Inc

2-Ethyl-6-(oxan-4-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223043-39-6
  • Molecular Weight : 317.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart