CO1H324A683A CombiPhos Catalysts Inc

2-(iso-Butyl)-6-(oxan-4-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223011-56-9
  • Molecular Weight : 345.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart