CO1H324A6845 CombiPhos Catalysts Inc

tert-butyl 3-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-yl)pyrrolidine-1-carboxylate

  • Cas Number : 2223003-71-0
  • Molecular Weight : 442.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart