CO1H324A684B CombiPhos Catalysts Inc

2-(2-methoxyphenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

  • Cas Number : 1402227-23-9
  • Molecular Weight : 312.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart