CO1H324A6865 CombiPhos Catalysts Inc

4-(1,4-Dioxan-2-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223006-80-0
  • Molecular Weight : 291.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart