CO1H324A6874 CombiPhos Catalysts Inc

2-(Oxolan-3-yl)-6-(1,4-dioxan-2-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223004-02-0
  • Molecular Weight : 361.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart