CO1H324A6879 CombiPhos Catalysts Inc

2-(N-Boc-Piperidin-4-yl)-6-(1,4-dioxan-2-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2222996-08-7
  • Molecular Weight : 474.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart