CO1H324A687C CombiPhos Catalysts Inc

2-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)piperazine

  • Cas Number : 2222996-21-4
  • Molecular Weight : 289.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart