CO1H324A6880 CombiPhos Catalysts Inc

tert-butyl 2-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine-4-carboxylate

  • Cas Number : 2222996-46-3
  • Molecular Weight : 390.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart