CO1H324A6888 CombiPhos Catalysts Inc

6'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-[1,2'-bipyridin]-2-one

  • Cas Number : 1352754-45-0
  • Molecular Weight : 298.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart