CO1H324A688E CombiPhos Catalysts Inc

2-(N-Boc-Pyrrolidin-3-yl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1350543-49-5
  • Molecular Weight : 375.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart