CO1H324A6892 CombiPhos Catalysts Inc

4-(4-Bromopyridin-2-yl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223012-87-9
  • Molecular Weight : 366.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart