CO1H324A6896 CombiPhos Catalysts Inc

4-(Pyridin-2-yl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223033-99-4
  • Molecular Weight : 271.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart