CO1H324A689F CombiPhos Catalysts Inc

3-(4-Fluoropyridin-2-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223049-50-9
  • Molecular Weight : 299.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart