CO1H324A68A0 CombiPhos Catalysts Inc

3-(6-Trifluoromethylpyridin-2-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223043-70-5
  • Molecular Weight : 349.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart