CO1H324A68A3 CombiPhos Catalysts Inc

3-(6-Bromopyridin-2-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223034-81-7
  • Molecular Weight : 360.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart