CO1H324A68AF CombiPhos Catalysts Inc

3-[4-(Methoxycarbonyl)pyridin-2-yl]phenylboronic acid pinacol ester

  • Cas Number : 2223049-26-9
  • Molecular Weight : 339.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart