CO1H324A68B0 CombiPhos Catalysts Inc

3-(Pyridin-2-yl)-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223046-24-8
  • Molecular Weight : 321.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart