CO1H324A68B5 CombiPhos Catalysts Inc

3-(Pyridin-2-yl)-5-chlorophenylboronic acid pinacol ester

  • Cas Number : 2223011-84-3
  • Molecular Weight : 315.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart