CO1H324A68B8 CombiPhos Catalysts Inc

2-(3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrazine

  • Cas Number : 2223052-61-5
  • Molecular Weight : 296.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart