CO1H324A68C7 CombiPhos Catalysts Inc

2-(Cyclohexyloxy)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1402174-34-8
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart