CO1H324A68CB CombiPhos Catalysts Inc

2-((tetrahydrofuran-3-yl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

  • Cas Number : 1353877-86-7
  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart