CO1H324A68CC CombiPhos Catalysts Inc

2,6-DI(N-BOC-PIPERIDIN-4-YL)PYRIDINE-4-BORONIC ACID PINACOL ESTER

  • Cas Number : 2223011-67-2
  • Molecular Weight : 571.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart