CO1H324A68D6 CombiPhos Catalysts Inc

5-(1-Hydroxy-1-methylethyl)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 2223035-79-6
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart