CO1H324A68D8 CombiPhos Catalysts Inc

2-(4-Tetrahydropyranyl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1402166-38-4
  • Molecular Weight : 290.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart