CO1H324A68DC CombiPhos Catalysts Inc

2-(3-Methylpiperidino)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1402174-20-2
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart