CO1H324A68E3 CombiPhos Catalysts Inc

2-(4-Hydroxypiperidin-1-yl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1202805-26-2
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart