CO1H324A68EC CombiPhos Catalysts Inc

2-(3-Cyanophenyl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 2223034-91-9
  • Molecular Weight : 307.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart