CO1H324A68EE CombiPhos Catalysts Inc

2-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)thiazole

  • Cas Number : 2223035-84-3
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart