CO1H324A68FD CombiPhos Catalysts Inc

2-(2-Aminopropan-2-yl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 2223049-96-3
  • Molecular Weight : 263.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart