CO1H324A6907 CombiPhos Catalysts Inc

3-(2-Aminopropan-2-yl)phenylboronic acid pinacol ester

  • Cas Number : 1075683-56-5
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart