CO1H324A6909 CombiPhos Catalysts Inc

2-Fluoro-6-(3-tolyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223012-40-4
  • Molecular Weight : 313.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart