CO1H324A690B CombiPhos Catalysts Inc

2-Fluoro-6-(n-propyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223050-03-9
  • Molecular Weight : 265.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart