CO1H324A6917 CombiPhos Catalysts Inc

2-Fluoro-6-(iso-butyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223050-11-9
  • Molecular Weight : 279.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart