CO1H324A691C CombiPhos Catalysts Inc

2-Fluoro-6-bromopyridine-3-boronic acid pinacol ester

  • Cas Number : 1310404-04-6
  • Molecular Weight : 301.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart