CO1H324A6921 CombiPhos Catalysts Inc

2-Fluoro-6-(2-fluorophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223037-30-5
  • Molecular Weight : 317.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart