CO1H324A6929 CombiPhos Catalysts Inc

2-Fluoro-6-(thiazol-5-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223036-00-6
  • Molecular Weight : 306.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart